Piperacillin sodium salt 98+%

Supplier: Ambeed
Danger

Synonyms: Piperacillin sodium, Sodium (2S,5R,6R)-6-((R)-2-(4-ethyl-2,3-dioxopiperazine-1-carboxamido)-2-phenylacetamido)-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate

A881033-1G A881033-250MG A881033-5G A881033-100MG A881033-25G
77603-206EA 20.5 USD
77603-206 77670-276 77670-278 77880-702 77880-703
Piperacillin sodium salt 98+%
Piperacillin sodium salt

Precaución: for Research Use Only

Formula: C₂₃H₂₇N₅O₇S
MW: 539.54 g/mol
MDL Number: MFCD00056867
CAS Number: 59703-84-3
UN: 2811
ADR: 6.1 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR