3-Bromoazetidine hydrochloride ≥98%, Ambeed.Inc

Supplier: Ambeed
Warning

A880645-1G A880645-5G A880645-10G
77684-845EA 48 USD
77684-845 77886-884 77886-883
3-Bromoazetidine hydrochloride ≥98%, Ambeed.Inc
3-Bromoazetidine hydrochloride

Precaución: For research use only.

Formula: C₃H₆BrN·ClH
MDL Number: MFCD09834130
CAS Number: 53913-82-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR