Maytansinol ≥98%

Supplier: Ambeed
Warning

Synonyms: (14S,16S,32S,33S,2R,4S,10E,12E,14R)-86-chloro-14,4-dihydroxy-85,14-dimethoxy-33,2,7,10-tetramethyl-7-aza-1(6,4)-oxazinana-3(2,3)-oxirana-8(1,3)-benzenacyclotetradecaphane-10,12-diene-12,6-dione, Ansamitocin P-0

A880207-10MG A880207-1MG A880207-5MG
77614-576EA 153.47 USD
77614-576 77614-578 77614-580
Maytansinol ≥98%
Maytansinol

Precaución: for Research Use Only

Formula: C₂₈H₃₇ClN₂O₈
MW: 565.06 g/mol
MDL Number: MFCD28144515
CAS Number: 57103-68-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR