N-Carbobenzoxy-4-hydroxy-D-2-phenylglycine 95+%

Supplier: Ambeed
Warning

A879599-5G A879599-100G A879599-25G
77674-312EA 14 USD
77674-312 77884-824 77884-825
N-Carbobenzoxy-4-hydroxy-D-2-phenylglycine 95+%
N-Carbobenzoxy-4-hydroxy-D-2-phenylglycine

Precaución: for Research Use Only

Formula: C₁₆H₁₅NO₅
MDL Number: MFCD09756650
CAS Number: 26787-75-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR