2-Bromo-3-(difluoromethyl)pyridine ≥98%

Supplier: Ambeed
Warning

A879198-1G A879198-250MG A879198-5G
77686-918EA 49 USD
77686-918 77901-623 77901-590
2-Bromo-3-(difluoromethyl)pyridine ≥98%
2-Bromo-3-(difluoromethyl)pyridine

Precaución: for Research Use Only

Formula: C₆H₄BrF₂N
MDL Number: MFCD18257230
CAS Number: 1211515-00-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR