Ethyl 2-(thietan-3-ylidene)acetate ≥98%

Supplier: Ambeed
Warning

A878783-250MG A878783-100MG
77749-684EA 44 USD
77749-684 77749-683
Ethyl 2-(thietan-3-ylidene)acetate ≥98%
Ethyl 2-(thietan-3-ylidene)acetate

Precaución: for Research Use Only

Formula: C₇H₁₀O₂S
MW: 158.218 g/mol
MDL Number: MFCD12755198
CAS Number: 1223573-30-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR