Nickel(II) phthalocyanine ≥98%

Supplier: Ambeed
Danger

A878313-1G A878313-5G A878313-25G A878313-10G
77603-194EA 12.3 USD
77603-194 77603-196 77670-274 77880-698
Nickel(II) phthalocyanine ≥98%
Nickel(II) phthalocyanine

Precaución: for Research Use Only

Formula: Ni(C₃₂H₁₆N₈)
MW: 571.22 g/mol
MDL Number: MFCD00041954
CAS Number: 14055-02-8
UN: 2811
ADR: 6.1 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR