4-(Trifluoromethyl)benzenesulfinic acid sodium salt ≥95%

Supplier: Ambeed
Warning

A876497-1G A876497-5G
77614-572EA 166.86 USD
77614-572 77901-132
4-(Trifluoromethyl)benzenesulfinic acid sodium salt ≥95%
4-(Trifluoromethyl)benzenesulfinic acid sodium salt

Precaución: for Research Use Only

Formula: C₇H₄F₃NaO₂S
MDL Number: MFCD11618070
CAS Number: 89520-63-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR