(S)-tert-Butyl 2-formylpiperidine-1-carboxylate ≥97%

Supplier: Ambeed
Warning

A876269-1G A876269-5G
77573-770EA 47.38 USD
77573-770 77684-841
(S)-tert-Butyl 2-formylpiperidine-1-carboxylate ≥97%
(S)-tert-Butyl 2-formylpiperidine-1-carboxylate

Precaución: For research use only.

Formula: C₁₁H₁₉NO₃
MW: 213.27 g/mol
MDL Number: MFCD06738713
CAS Number: 150521-32-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR