2-Benzyl-6,8-diphenylimidazo[1,2-a]pyrazin-3(7H)-one ≥97%

Supplier: Ambeed
Warning

Synonyms: Diphenylterazine

A875856-1MG A875856-5MG A875856-10MG
77599-204EA 109.68 USD
77599-204 77599-206 77599-202
2-Benzyl-6,8-diphenylimidazo[1,2-a]pyrazin-3(7H)-one ≥97%
2-Benzyl-6,8-diphenylimidazo[1,2-a]pyrazin-3(7H)-one

Precaución: for Research Use Only

Formula: C₂₅H₁₉N₃O
MDL Number: MFCD31619283
CAS Number: 344940-63-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR