(3-(2-(Piperidin-1-yl)ethoxy)phenyl)boronic acid ≥98%

Supplier: Ambeed
Warning

A874029-250MG A874029-100MG
77784-301EA 228 USD
77784-301 77784-300
(3-(2-(Piperidin-1-yl)ethoxy)phenyl)boronic acid ≥98%
(3-(2-(Piperidin-1-yl)ethoxy)phenyl)boronic acid

Precaución: for Research Use Only

Formula: C₁₃H₂₀BNO₃
MDL Number: MFCD11540656
CAS Number: 1311166-07-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR