3-Bromo-5-cyanobenzoic acid methyl ester ≥98%

Supplier: Ambeed
Danger

A873610-100MG A873610-250MG
77884-819EA 14 USD
77884-819 77884-820
3-Bromo-5-cyanobenzoic acid methyl ester ≥98%
3-Bromo-5-cyanobenzoic acid methyl ester

Precaución: for Research Use Only

Formula: C₉H₆BrNO₂
MDL Number: MFCD16620379
CAS Number: 453566-15-9
UN: 3439
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR