5,6-Dihydro-imidazo[4,5,l-j-k]-[1]benzazepine-2,7-[1H,4H]-dione ≥98%
Supplier: Ambeed
|
Warning
|
A873398-1G
A873398-5G
77893-674EA
85
USD
77893-674
77893-675
5,6-Dihydro-imidazo[4,5,l-j-k]-[1]benzazepine-2,7-[1H,4H]-dione ≥98%
5,6-Dihydro-imidazo[4,5,l-j-k]-[1]benzazepine-2,7-[1H,4H]-dione
Precaución: for Research Use Only
Formula:
C₁₁H₁₀N₂O₂ |
MDL Number:
MFCD18252621 CAS Number: 92260-81-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...