1-(4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)cyclobutanol ≥95%
Supplier: Ambeed
|
Warning
|
A871253-100MG
A871253-1G
77684-835EA
114
USD
77684-835
77684-836
1-(4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)cyclobutanol ≥95%
1-(4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)cyclobutanol
Precaución: For research use only.
Formula:
C₁₆H₂₃BO₃ MW: 274.163 g/mol |
MDL Number:
MFCD18734557 CAS Number: 1398331-98-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...