1-(4-Iodophenyl)-2,5-dimethylpyrrole ≥98%

Supplier: Ambeed
Warning

Synonyms: 1-(4-Iodophenyl)-2,5-dimethyl-1H-pyrrole

A871192-250MG A871192-1G
77595-406EA 18.45 USD
77595-406 77674-298
1-(4-Iodophenyl)-2,5-dimethylpyrrole ≥98%
1-(4-Iodophenyl)-2,5-dimethylpyrrole

Precaución: for Research Use Only

Formula: C₁₂H₁₂IN
MDL Number: MFCD02692647
CAS Number: 288608-09-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR