Cobalt(II) phosphate ≥98%

Supplier: Ambeed
Warning

A870676-5G A870676-100G A870676-25G
77627-694EA 22.66 USD
77627-694 77880-690 77880-691
Cobalt(II) phosphate ≥98%
Cobalt(II) phosphate

Precaución: for Research Use Only

Formula: Co₃O₈P₂
MDL Number: MFCD00016028
CAS Number: 13455-36-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR