5-Methyl-1H-indazole-4-carboxylic acid ≥97%

Supplier: Ambeed
Warning

A869002-100MG A869002-250MG
77623-082EA 193.64 USD
77623-082 77623-084
5-Methyl-1H-indazole-4-carboxylic acid ≥97%
5-Methyl-1H-indazole-4-carboxylic acid

Precaución: for Research Use Only

Formula: C₉H₈N₂O₂
MDL Number: MFCD22556693
CAS Number: 1360927-57-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR