1-(Benzyloxy)-2-iodobenzene ≥98%

Supplier: Ambeed
Warning

A868751-1G A868751-250MG A868751-100MG
77784-274EA 93 USD
77784-274 77784-275 77784-273
1-(Benzyloxy)-2-iodobenzene ≥98%
1-(Benzyloxy)-2-iodobenzene

Precaución: for Research Use Only

Formula: C₁₃H₁₁IO
MDL Number: MFCD14648490
CAS Number: 142523-69-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR