Pentaethylene glycol monobenzyl ether ≥95%
Supplier: Ambeed
|
Warning
|
Synonyms:
BnO-PEG5-OH, 2-[2-[2-[2-[2-(Benzyloxy)ethoxy]ethoxy]ethoxy]ethoxy]ethanol, 1-Phenyl-2,5,8,11,14-pentaoxahexadecan-16-ol
A868571-1G
A868571-10G
A868571-5G
77614-566EA
20.6
USD
77614-566
77684-831
77684-832
Pentaethylene glycol monobenzyl ether ≥95%
Pentaethylene glycol monobenzyl ether
Precaución: For research use only.
Formula:
C₁₇H₂₈O₆ MW: 328.41 g/mol |
MDL Number:
MFCD06797154 CAS Number: 57671-28-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...