Ensartinib ≥99%

Supplier: Ambeed
Warning

Synonyms: X-376, (R)-6-Amino-5-(1-(2,6-dichloro-3-fluorophenyl)ethoxy)-N-(4-(4-methylpiperazine-1-carbonyl)phenyl)pyridazine-3-carboxamide

A868332-100MG A868332-250MG
77747-639EA 103 USD
77747-639 77747-640
Ensartinib ≥99%
Ensartinib

Precaución: for Research Use Only

Formula: C₂₅H₂₅Cl₂FN₆O₃
MW: 547.41 g/mol
MDL Number: MFCD25976868
CAS Number: 1365267-27-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR