Iron(III) 2-hydroxypropane-1,2,3-tricarboxylate AR

Supplier: Ambeed
Warning

Synonyms: Ferric citrate

A868163-100G A868163-25G A868163-500G
77595-396EA 19.48 USD
77595-396 77595-398 77595-400
Iron(III) 2-hydroxypropane-1,2,3-tricarboxylate AR
Iron(III) 2-hydroxypropane-1,2,3-tricarboxylate

Precaución: for Research Use Only

Formula: C₆H₅FeO₇
MW: 244.94 g/mol
MDL Number: MFCD00149617
CAS Number: 3522-50-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR