1,5-Dibromo-2,4-diiodobenzene ≥98%

Supplier: Ambeed
Danger

A868066-1G A868066-250MG A868066-5G
77623-078EA 90.64 USD
77623-078 77784-268 77784-269
1,5-Dibromo-2,4-diiodobenzene ≥98%
1,5-Dibromo-2,4-diiodobenzene

Precaución: for Research Use Only

Formula: C₆H₂Br₂I₂
MDL Number: MFCD00192720
CAS Number: 96843-23-1
UN: 1759
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR