1-Methylcyclobutan-1-ol ≥95%

Supplier: Ambeed
Danger

A867930-1G A867930-100MG A867930-250MG A867930-5G
77674-290EA 83 USD
77674-290 77884-806 77884-807 77884-808
1-Methylcyclobutan-1-ol ≥95%
1-Methylcyclobutan-1-ol

Precaución: for Research Use Only

Formula: C₅H₁₀O
MDL Number: MFCD01218890
CAS Number: 20117-47-9
UN: 1993
ADR: 3,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR