2-Methyl-5-isopropylaniline ≥97%
Supplier: Ambeed
|
Warning
|
A867813-1G
A867813-25G
A867813-5G
77627-688EA
17.1
USD
77627-688
77878-708
77878-709
2-Methyl-5-isopropylaniline ≥97%
2-Methyl-5-isopropylaniline
Precaución: for Research Use Only
Formula:
C₁₀H₁₅N |
MDL Number:
MFCD00044503 CAS Number: 2051-53-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...