(14S,16S,32S,33S,2R,4S,10E,12E,14R)-86-Chloro-14-hydroxy-85,14-dimethoxy-33,2,7,10-tetramethyl-12,6-dioxo-7-aza-1(6,4)-oxazinana-3(2,3)-oxirana-8(1,3)-benzenacyclotetradecaphane-10,12-dien-4-yl N-methyl-N-(3-(methyldisulfaneyl)propanoyl)-L-alaninate ≥98%

Supplier: Ambeed
Danger

Synonyms: DM1-SMe

A866930-10MG A866930-1MG A866930-5MG
77618-438EA 157.59 USD
77618-438 77618-440 77618-442
(14S,16S,32S,33S,2R,4S,10E,12E,14R)-86-Chloro-14-hydroxy-85,14-dimethoxy-33,2,7,10-tetramethyl-12,6-dioxo-7-aza-1(6,4)-oxazinana-3(2,3)-oxirana-8(1,3)-benzenacyclotetradecaphane-10,12-dien-4-yl N-methyl-N-(3-(methyldisulfaneyl)propanoyl)-L-alaninate ≥98%
(14S,16S,32S,33S,2R,4S,10E,12E,14R)-86-Chloro-14-hydroxy-85,14-dimethoxy-33,2,7,10-tetramethyl-12,6-dioxo-7-aza-1(6,4)-oxazinana-3(2,3)-oxirana-8(1,3)-benzenacyclotetradecaphane-10,12-dien-4-yl N-methyl-N-(3-(methyldisulfaneyl)propanoyl)-L-alaninate

Precaución: for Research Use Only

Formula: C₃₆H₅₀ClN₃O₁₀S₂
MW: 784.37 g/mol
MDL Number: MFCD28400100
CAS Number: 138148-68-2
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR