(5-Chloro-3-fluoropyridin-2-yl)methanol ≥98%, Ambeed.Inc

Supplier: Ambeed
Warning

A865989-1G A865989-100MG A865989-250MG
77684-827EA 229 USD
77684-827 77891-628 77891-629
(5-Chloro-3-fluoropyridin-2-yl)methanol ≥98%, Ambeed.Inc
(5-Chloro-3-fluoropyridin-2-yl)methanol

Precaución: For research use only.

Formula: C₆H₅ClFNO
MW: 161.561 g/mol
MDL Number: MFCD16607027
CAS Number: 214055-12-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR