1-(2-Thiazolyl)ethylamine ≥95%

Supplier: Ambeed
Danger

A865894-250MG A865894-1G
77674-288EA 16 USD
77674-288 77674-286
1-(2-Thiazolyl)ethylamine ≥95%
1-(2-Thiazolyl)ethylamine

Precaución: for Research Use Only

Formula: C₅H₈N₂S
MW: 128.20 g/mol
MDL Number: MFCD02854206
CAS Number: 432047-36-4
UN: 2735
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR