4'-Chloro-3'-fluoroacetophenone ≥98%

Supplier: Ambeed
Warning

Synonyms: 1-(4-Chloro-3-fluorophenyl)ethanone

A865856-1G A865856-5G
77573-748EA 11.33 USD
77573-748 77747-634
4'-Chloro-3'-fluoroacetophenone ≥98%
4'-Chloro-3'-fluoroacetophenone

Precaución: for Research Use Only

Formula: C₈H₆ClFO
MW: 172.59 g/mol
MDL Number: MFCD04115858
CAS Number: 151945-84-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR