1-Palmitoyl-sn-glycero-3-phosphocholine ≥98%

Supplier: Ambeed
Warning

Synonyms: 1-Palmitoyl-sn-glycero-3-phosphocholine, (R)-2-Hydroxy-3-(palmitoyloxy)propyl (2-(trimethylammonio)ethyl) phosphate

A865426-100MG A865426-1G A865426-250MG
77603-176EA 55.35 USD
77603-176 77614-556 77686-905
1-Palmitoyl-sn-glycero-3-phosphocholine ≥98%
1-Palmitoyl-sn-glycero-3-phosphocholine

Precaución: for Research Use Only

Formula: C₂₄H₅₀NO₇P
MW: 495.638 g/mol
MDL Number: MFCD00036904
CAS Number: 17364-16-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR