Methyl 2-nitro-4-(trifluoromethyl)benzoate ≥95%
Supplier: Ambeed
|
Warning
|
A865013-1G
A865013-5G
77891-625EA
12
USD
77891-625
77891-626
Methyl 2-nitro-4-(trifluoromethyl)benzoate ≥95%
Methyl 2-nitro-4-(trifluoromethyl)benzoate
Precaución: for Research Use Only
Formula:
C₉H₆F₃NO₄ |
MDL Number:
MFCD08282772 CAS Number: 228418-45-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...