7-Chloronaphthalen-1-amine ≥95%
Supplier: Ambeed
|
Warning
|
A864460-250MG
A864460-1G
77614-552EA
90.64
USD
77614-552
77684-826
7-Chloronaphthalen-1-amine ≥95%
7-Chloronaphthalen-1-amine
Precaución: For research use only.
Formula:
C₁₀H₈ClN MW: 177.63 g/mol |
MDL Number:
MFCD17012303 CAS Number: 50987-58-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...