1-(4,4-Diphenylbut-3-en-1-yl)piperidine-3-carboxylic acid hydrochloride ≥95%

Supplier: Ambeed
Warning

Synonyms: SKF89976A HCl

A864319-5MG A864319-10MG A864319-100MG
77747-632EA 63 USD
77747-632 77747-631 77747-630
1-(4,4-Diphenylbut-3-en-1-yl)piperidine-3-carboxylic acid hydrochloride ≥95%
1-(4,4-Diphenylbut-3-en-1-yl)piperidine-3-carboxylic acid hydrochloride

Precaución: for Research Use Only

Formula: C₂₂H₂₆ClNO₂
MW: 371.9 g/mol
MDL Number: MFCD01321071
CAS Number: 85375-15-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR