tert-Butyl N-[6-(methylamino)hexyl]carbamate ≥95%
Supplier: Ambeed
|
Danger
|
A863884-100MG
A863884-1G
A863884-250MG
A863884-5G
77599-186EA
46.13
USD
77599-186
77599-188
77599-190
77599-192
tert-Butyl N-[6-(methylamino)hexyl]carbamate ≥95%
tert-Butyl N-[6-(methylamino)hexyl]carbamate
Precaución: for Research Use Only
Formula:
C₁₂H₂₆N₂O₂ |
MDL Number:
MFCD30535604 CAS Number: 1013915-06-4 UN: 2735 ADR: 8,Ⅲ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...