Spiro[3.5]nonan-7-one ≥95%

Supplier: Ambeed
Warning

A863757-1G A863757-250MG
77886-858EA 652 USD
77886-858 77886-859
Spiro[3.5]nonan-7-one ≥95%
Spiro[3.5]nonan-7-one

Precaución: for Research Use Only

Formula: C₉H₁₄O
MDL Number: MFCD17012716
CAS Number: 31058-02-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR