1-(3,4-Dimethoxyphenyl)ethylamine ≥95%

Supplier: Ambeed
Danger

A863024-100MG A863024-250MG
77873-324EA 33 USD
77873-324 77873-325
1-(3,4-Dimethoxyphenyl)ethylamine ≥95%
1-(3,4-Dimethoxyphenyl)ethylamine

Precaución: for Research Use Only

Formula: C₁₀H₁₅NO₂
MDL Number: MFCD00561923
CAS Number: 50919-08-9
UN: 2735
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR