1-(2-Tetrahydropyranyl)-1H-pyrazole-5-boronic acid ≥98%
Supplier: Ambeed
|
Warning
|
A862846-100MG
A862846-1G
77886-856EA
97
USD
77886-856
77886-857
1-(2-Tetrahydropyranyl)-1H-pyrazole-5-boronic acid ≥98%
1-(2-Tetrahydropyranyl)-1H-pyrazole-5-boronic acid
Precaución: for Research Use Only
Formula:
C₈H₁₃BN₂O₃ |
MDL Number:
MFCD11046121 CAS Number: 1105511-68-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...