6-(4,4,5,5-Tetramethyl1,3,2-dioxaboralan-2-yl)-2,3-dihydroinden-1-one ≥98%

Supplier: Ambeed
Warning

Synonyms: 6-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-2,3-dihydro-1H-inden-1-one

A862594-100MG A862594-1G A862594-250MG
77747-627EA 67 USD
77747-627 77747-628 77747-629
6-(4,4,5,5-Tetramethyl1,3,2-dioxaboralan-2-yl)-2,3-dihydroinden-1-one ≥98%
6-(4,4,5,5-Tetramethyl1,3,2-dioxaboralan-2-yl)-2,3-dihydroinden-1-one

Precaución: for Research Use Only

Formula: C₁₅H₁₉BO₃
MW: 258.121 g/mol
MDL Number: MFCD08706296
CAS Number: 915402-06-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR