Cefaclor monohydrate ≥99%

Supplier: Ambeed
Warning

A862563-1G A862563-1MG A862563-250MG A862563-5G
77603-168EA 19.48 USD
77603-168 77627-678 77627-680 77627-682
Cefaclor monohydrate ≥99%
Cefaclor monohydrate
Formula: C₁₅H₁₄ClN₃O₄S·H₂O
MDL Number: MFCD00071999
CAS Number: 70356-03-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR