Difluoromethyl phenyl sulfide ≥98% (by GC)

Supplier: Ambeed
Danger

Synonyms: alpha,alpha-Difluorothioanisole, Difluoromethyl Phenyl sulphide, (Difluoromethylthio)benzene

A862472-1G A862472-5G A862472-250MG
77623-060EA 62.83 USD
77623-060 77623-062 77784-257
Difluoromethyl phenyl sulfide ≥98% (by GC)
Difluoromethyl phenyl sulfide

Precaución: for Research Use Only

Formula: C₇H₆F₂S
MDL Number: MFCD16301013
CAS Number: 1535-67-7
UN: 1993
ADR: 3,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR