(S)-2-((((S)-3-(4-Chlorophenyl)-4-phenyl-4,5-dihydro-1H-pyrazol-1-yl)(4-chlorophenylsulfonamido)methylene)amino)-3-methylbutanamide ≥98%
Supplier: Ambeed
|
Warning
|
Synonyms:
JD-5037
A862077-10MG
A862077-1MG
A862077-5MG
77618-424EA
134.93
USD
77618-424
77618-426
77618-428
(S)-2-((((S)-3-(4-Chlorophenyl)-4-phenyl-4,5-dihydro-1H-pyrazol-1-yl)(4-chlorophenylsulfonamido)methylene)amino)-3-methylbutanamide ≥98%
(S)-2-((((S)-3-(4-Chlorophenyl)-4-phenyl-4,5-dihydro-1H-pyrazol-1-yl)(4-chlorophenylsulfonamido)methylene)amino)-3-methylbutanamide
Precaución: for Research Use Only
Formula:
C₂₇H₂₇Cl₂N₅O₃S MW: 572.51 g/mol |
MDL Number:
MFCD30533451 CAS Number: 1392116-14-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...