Ethyl hippurate 95+%

Supplier: Ambeed
Warning

A861984-100G A861984-1G A861984-25G A861984-5G
77880-672EA 112 USD
77880-672 77880-673 77880-674 77880-675
Ethyl hippurate 95+%
Ethyl hippurate

Precaución: for Research Use Only

Formula: C₁₁H₁₃NO₃
MDL Number: MFCD00026890
CAS Number: 1499-53-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR