8-Chloro-6-methyl-[1,3]dioxolo[4,5-g]quinoline 95+%, Ambeed.Inc

Supplier: Ambeed
Danger

A861915-1G A861915-250MG
77684-824EA 133 USD
77684-824 77891-623
8-Chloro-6-methyl-[1,3]dioxolo[4,5-g]quinoline 95+%, Ambeed.Inc
8-Chloro-6-methyl-[1,3]dioxolo[4,5-g]quinoline

Precaución: For research use only.

Formula: C₁₁H₈ClNO₂
MDL Number: MFCD11868303
CAS Number: 50593-65-2
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR