1H-Benzimidazole-6-boronic acid ≥98%

Supplier: Ambeed
Warning

Synonyms: (1H-Benzo[d]imidazol-5-yl)boronic acid

A860814-250MG A860814-100MG A860814-1G A860814-5G
77573-738EA 110 USD
77573-738 77674-274 77884-794 77884-795
1H-Benzimidazole-6-boronic acid ≥98%
1H-Benzimidazole-6-boronic acid

Precaución: for Research Use Only

Formula: C₇H₇BN₂O₂
MW: 161.95 g/mol
MDL Number: MFCD18383659
CAS Number: 1228183-22-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR