4,5-Dimethoxy-2-methylaniline ≥95%

Supplier: Ambeed
Warning

A860778-1G A860778-5G
77627-672EA 14.42 USD
77627-672 77627-674
4,5-Dimethoxy-2-methylaniline ≥95%
4,5-Dimethoxy-2-methylaniline

Precaución: for Research Use Only

Formula: C₉H₁₃NO₂
MW: 167.210 g/mol
MDL Number: MFCD00130067
CAS Number: 41864-45-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR