N-(5-(4-Bromophenyl)-6-(2-((5-bromopyrimidin-2-yl)oxy)ethoxy)pyrimidin-4-yl)sulfamide ≥98%

Supplier: Ambeed
Warning

Synonyms: Aprocitentan

A860741-10MG A860741-1MG A860741-5MG
77618-416EA 286.34 USD
77618-416 77618-418 77618-420
N-(5-(4-Bromophenyl)-6-(2-((5-bromopyrimidin-2-yl)oxy)ethoxy)pyrimidin-4-yl)sulfamide ≥98%
N-(5-(4-Bromophenyl)-6-(2-((5-bromopyrimidin-2-yl)oxy)ethoxy)pyrimidin-4-yl)sulfamide

Precaución: for Research Use Only

Formula: C₁₆H₁₄Br₂N₆O₄S
MW: 546.19 g/mol
MDL Number: MFCD17167076
CAS Number: 1103522-45-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR