3-Methylbicyclo[1.1.1]pentane-1-carboxylic acid ≥98%

Supplier: Ambeed
Warning

A860178-250MG A860178-100MG
77573-736EA 225.57 USD
77573-736 77749-667
3-Methylbicyclo[1.1.1]pentane-1-carboxylic acid ≥98%
3-Methylbicyclo[1.1.1]pentane-1-carboxylic acid

Precaución: for Research Use Only

Formula: C₇H₁₀O₂
MW: 126.153 g/mol
MDL Number: MFCD19228632
CAS Number: 65862-01-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR