7-Methoxy-4-quinolinol ≥98%

Supplier: Ambeed
Danger

Synonyms: 4-Hydroxy-7-methoxyquinoline

A859565-1G A859565-5G
77880-666EA 20 USD
77880-666 77880-667
7-Methoxy-4-quinolinol ≥98%
7-Methoxy-4-quinolinol

Precaución: for Research Use Only

Formula: C₁₀H₉NO₂
MDL Number: MFCD00169015
CAS Number: 82121-05-9
UN: 1759
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR