2-Bromo-6-iodophenol ≥95%

Supplier: Ambeed
Danger

A859175-1G A859175-5G
77614-540EA 52.53 USD
77614-540 77884-792
2-Bromo-6-iodophenol ≥95%
2-Bromo-6-iodophenol

Precaución: for Research Use Only

Formula: C₆H₄BrIO
MDL Number: MFCD16999870
CAS Number: 2040-86-0
UN: 1759
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR