2-Cyanoethyl 3-Oxobutyrate ≥95%, Ambeed.Inc

Supplier: Ambeed
Warning

A858873-5G A858873-25G
77684-818EA 72 USD
77684-818 77893-663
2-Cyanoethyl 3-Oxobutyrate ≥95%, Ambeed.Inc
2-Cyanoethyl 3-Oxobutyrate

Precaución: For research use only.

Formula: C₇H₉NO₃
MW: 155.15 g/mol
MDL Number: MFCD24687999
CAS Number: 65193-87-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR