4-Bromo-6-fluoropyrimidine ≥95%

Supplier: Ambeed
Warning

A858564-5G A858564-1G A858564-250MG
77573-728EA 1055 USD
77573-728 77891-621 77891-622
4-Bromo-6-fluoropyrimidine ≥95%
4-Bromo-6-fluoropyrimidine

Precaución: for Research Use Only

Formula: C₄H₂BrFN₂
MDL Number: MFCD14702771
CAS Number: 1209458-29-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR